Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025052.1 | NP_001020223.1 | 1 | 1707 | ||
GenBank mRNA | BC079199.1 | - | 1 | 1707 | 7131579 |