Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278610.1 | NP_001265539.1 | 2 | 350 | ||
NM_002466.3 | NP_002457.1 | 2 | 350 | |||
GenBank mRNA | AK223482.1 | - | 2 | 85 | ||
AK303249.1 | - | 2 | 255 | |||
AK314791.1 | - | 2 | 255 | |||
BC007585.1 | - | 2 | 275 | 3162656 | ||
BC053555.1 | - | 2 | 244 | 6140320 | ||
BU178833.1 | - | 300 | ||||
X13293.1 | - | 2 | 212 |