Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mesp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008588.2 | NP_032614.2 | 2 | 953 | ||
GenBank mRNA | AK006394.1 | - | 2 | 384 | ||
BC012689.1 | - | 2 | 1162 | |||
CF105615.1 | - | 2 | 428 | |||
D83674.1 | - | 2 | 1016 |