Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252454.1 | NP_001239383.1 | 11 | 2070 | ||
NM_010147.4 | NP_034277.1 | 12 | 2241 | |||
GenBank mRNA | AF057285.1 | - | 4 | 714 | ||
AK155033.1 | - | 12 | 2237 | |||
BC046962.1 | - | 10 | 1876 | 6391968 | ||
BC067206.1 | - | 11 | 2012 | 6514327 | ||
BC099682.1 | - | 12 | 2171 | 6529672 |