Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011836.3 | NP_035966.2 | 28 | 5645 | ||
GenBank mRNA | AK132948.1 | - | 25 | 4632 | ||
BC005535.1 | - | 11 | 2390 | |||
BC022578.1 | - | 5 | 1613 | |||
BC096366.1 | - | 28 | 5645 | 2865309 |