Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIH1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017916.2 | NP_060386.1 | 3 | 479 | ||
XM_006723254.2 | XP_006723317.1 | 2 | 105 | |||
GenBank mRNA | AK000650.1 | - | 3 | 431 | ||
AK294687.1 | - | 3 | 405 | |||
AK304476.1 | - | 3 | 530 | |||
BC001108.2 | - | 3 | 409 | 2966990 | ||
DC385084.1 | - | 3 | 479 |