Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AF251705 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134158.1 | NP_598919.1 | 4 | 1081 | ||
GenBank mRNA | AA175302.1 | - | 3 | 450 | ||
AB091767.1 | - | 4 | 812 | |||
AB091768.1 | - | 4 | 801 | |||
AF251705.1 | - | 4 | 1168 | |||
AK050008.1 | - | 4 | 809 | |||
AK079395.1 | - | 4 | 897 | |||
AY048685.1 | - | 4 | 1081 | |||
BC006801.1 | - | 4 | 812 | 3152857 |