Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URGCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077663.2 | NP_001071131.1 | 5 | 679 | ||
NM_001077664.2 | NP_001071132.1 | 5 | 451 | |||
NM_001290075.1 | NP_001277004.1 | 5 | 674 | |||
NM_001290076.1 | NP_001277005.1 | 6 | 573 | |||
NM_017920.4 | NP_060390.3 | 4 | 652 | |||
GenBank mRNA | AB040940.1 | - | 5 | 857 | ||
AK302459.1 | - | 6 | 573 | |||
AK314339.1 | - | 4 | 198 | |||
AL832134.1 | - | 3 | 2292 | |||
AY078404.1 | - | 4 | 171 | |||
BC018426.1 | - | 4 | 173 | 4213413 | ||
BG722670.1 | - | 5 | 429 | |||
BX640797.1 | - | 5 | 214 | |||
BX648505.1 | - | 5 | 219 | |||
CD628752.1 | - | 5 | 445 | |||
CR749398.1 | - | 5 | 231 | |||
DA240479.1 | - | 4 | 217 | |||
DA316326.1 | - | 5 | 235 | |||
DC336174.1 | - | 5 | 451 |
Gene Symbol: URGCP-MRPS24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204871.1 | NP_001191800.1 | 4 | 215 | ||
GenBank mRNA | AI027170.1 | - | 4 | 171 | ||
FY210844.1 | - | 3 | 123 |