Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181752.1 | NP_861417.1 | 1 | 1133 | ||
GenBank mRNA | AK132237.1 | - | 1269 | |||
AK141053.1 | - | 1 | 1217 | |||
AY288425.1 | - | 1 | 1133 | |||
BC062104.1 | - | 1166 | 6809932 | |||
BC114990.1 | - | 1 | 1132 | |||
BC115824.1 | - | 1 | 1132 | |||
BC139332.1 | - | 1 | 1195 | 8862354 | ||
BC139335.1 | - | 1 | 1195 | 8862357 | ||
BC148305.2 | - | 1 | 1133 |
Set Membership: |
G-protein coupled receptor |