Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam189a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183087.4 | NP_898910.2 | 9, 10 | 1432 | ||
GenBank mRNA | AK017751.1 | - | 3, 4 | 538 | ||
AK138167.1 | - | 7, 8 | 927 | |||
AK172974.1 | - | 9, 10 | 1204 | |||
BC049085.1 | - | 6, 7 | 754 | |||
BC147145.1 | - | 1, 2 | 146 | |||
BC147146.1 | - | 1, 2 | 146 |