Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dscc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183089.2 | NP_898912.2 | 2 | 318 | ||
GenBank mRNA | AK008137.1 | - | 2 | 319 | ||
AK011162.1 | - | 2 | 334 | |||
AK144502.1 | - | 2 | 321 | |||
BC116833.1 | - | 2 | 293 | 40126152 | ||
BC116835.1 | - | 2 | 293 | 40126154 |