Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013438.2 | NP_001013456.1 | 3 | 371 | ||
NM_001013439.2 | NP_001013457.1 | 4 | 715 | |||
NM_005087.3 | NP_005078.2 | 3 | 371 | |||
XM_005247813.3 | XP_005247870.2 | 3 | 146 | |||
XM_005247814.3 | XP_005247871.2 | 3 | 146 | |||
XM_005247815.3 | XP_005247872.2 | 3 | 146 | |||
XM_005247816.3 | XP_005247873.2 | 3 | 146 | |||
XM_006713775.3 | XP_006713838.1 | 4 | 489 | |||
XM_017007298.1 | XP_016862787.1 | 4 | 489 | |||
XM_017007299.1 | XP_016862788.1 | 4 | 489 | |||
XM_017007300.1 | XP_016862789.1 | 4 | 489 | |||
XM_017007301.1 | XP_016862790.1 | 4 | 489 | |||
GenBank mRNA | AK225334.1 | - | 3 | 147 | ||
AK292633.1 | - | 3 | 147 | |||
AK294603.1 | - | 2 | 93 | |||
AK302197.1 | - | 3 | 150 | |||
AU280585.1 | - | 4 | 490 | |||
AY341428.1 | - | 3 | 422 | |||
BC028983.1 | - | 3 | 141 | 5122574 | ||
DC352767.1 | - | 3 | 371 | |||
KR259877.1 | - | 3 | 131 | |||
KR259878.1 | - | 3 | 131 | |||
KR259879.1 | - | 3 | 131 | |||
KR259880.1 | - | 4 | 475 | |||
U25165.1 | - | 3 | 143 |