Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smim5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183259.3 | NP_899082.1 | 3 | 614 | ||
GenBank mRNA | AK004370.1 | - | 3 | 547 | ||
AK020450.1 | - | 3 | 576 | |||
BC065414.1 | - | 4 | 602 |