Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox6b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289848.1 | NP_001276777.1 | 2 | 269 | ||
NM_001289849.1 | NP_001276778.1 | 2 | 238 | |||
NM_001289850.1 | NP_001276779.1 | 2 | 238 | |||
NM_183405.2 | NP_899664.1 | 2 | 238 | |||
NM_183406.3 | NP_899665.1 | 1 | 149 | |||
GenBank mRNA | AK160152.1 | - | 2 | 168 | ||
AY152400.1 | - | 2 | 238 | |||
BC048670.1 | - | 1 | 149 | 6773974 | ||
BC107296.1 | - | 2 | 195 | 40053107 | ||
BC107297.1 | - | 2 | 195 | 40053112 | ||
BC125562.1 | - | 2 | 178 | 40130077 | ||
BC125564.1 | - | 2 | 181 | 40130079 | ||
BC145201.1 | - | 2 | 213 | 9053752 | ||
BG080371.2 | - | 2 | 146 | |||
CT010366.1 | - | 1 | 82 | |||
W88135.1 | - | 1 | 226 |