Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uts2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198166.3 | NP_937809.1 | 2 | 216 | ||
GenBank mRNA | AB116020.1 | - | 2 | 163 | ||
AK131972.1 | - | 2 | 194 | |||
AK131973.1 | - | 2 | 197 | |||
AK132025.1 | - | 2 | 217 | |||
AV144685.2 | - | 2 | 230 | |||
AY321315.1 | - | 2 | 197 | |||
BB611261.1 | - | 2 | 217 | |||
BC117024.1 | - | 2 | 188 | 40126343 | ||
BC117026.1 | - | 2 | 188 | 40126345 |