Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hbb-b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198776.1 | NP_942071.1 | 1 | 116 | ||
XM_006229913.2 | XP_006229975.2 | 1 | 150 | |||
GenBank mRNA | BC058502.1 | - | 1 | 116 | 6887460 | |
FQ218177.1 | - | 1 | 110 |
Gene Symbol: LOC103694855 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773799.1 | XP_008772021.1 | 1 | 152 | ||
XM_008773800.1 | XP_008772022.1 | 1 | 146 |