Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd300c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199225.1 | NP_954695.1 | 2 | 340 | ||
GenBank mRNA | AY457052.1 | - | 2 | 340 |