Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Laptm4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199384.2 | NP_955416.1 | 7 | 1194 | ||
GenBank mRNA | BC063179.1 | - | 7 | 1198 | 6890564 | |
FQ212732.1 | - | 7 | 1597 | |||
FQ213363.1 | - | 7 | 1228 | |||
FQ229677.1 | - | 1153 | ||||
FQ232171.1 | - | 7 | 1252 | |||
FQ232930.1 | - | 7 | 1212 | |||
FQ235113.1 | - | 7 | 1099 |