Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100a7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199422.1 | NP_955454.1 | 2 | 217 | ||
GenBank mRNA | AY465109.1 | - | 2 | 217 | ||
AY582964.1 | - | 1 | 124 | |||
BC107307.1 | - | 2 | 199 | 40053247 | ||
BC107308.1 | - | 2 | 199 | 40053248 |