Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201355.3 | NP_958743.1 | 3 | 1204 | ||
GenBank mRNA | AK078272.1 | - | 3 | 870 | ||
AK158627.1 | - | 3 | 1205 | |||
AK163549.1 | - | 3 | 934 | |||
BC047219.1 | - | 3 | 840 | 6491365 |