Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM45A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303111.1 | NP_001290040.1 | 10 | 2055 | ||
NM_001303112.1 | NP_001290041.1 | 9 | 1802 | |||
NM_001303113.1 | NP_001290042.1 | 8 | 1721 | |||
NM_207009.3 | NP_996892.1 | 9 | 1850 | |||
NR_130122.1 | 8 | 1738 | ||||
XM_006717849.3 | XP_006717912.1 | 9 | 1870 | |||
XM_006717850.3 | XP_006717913.1 | 10 | 2112 | |||
XM_006717851.3 | XP_006717914.1 | 10 | 2092 | |||
XM_006717852.3 | XP_006717915.1 | 10 | 2076 | |||
XM_017016261.1 | XP_016871750.1 | 10 | 2585 | |||
XM_017016262.1 | XP_016871751.1 | 11 | 2300 | |||
XM_017016263.1 | XP_016871752.1 | 11 | 2281 | |||
XM_017016264.1 | XP_016871753.1 | 9 | 1245 | |||
XM_017016265.1 | XP_016871754.1 | 10 | 2096 | |||
XM_017016266.1 | XP_016871755.1 | 11 | 2277 | |||
XM_017016267.1 | XP_016871756.1 | 11 | 2261 | |||
GenBank mRNA | AK027029.1 | - | 7 | 1950 |
Gene Symbol: FAM45BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027141.2 | 1 | 1988 |