Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282659.1 | NP_001269588.1 | 3 | 1069 | ||
NM_017944.3 | NP_060414.3 | 2 | 865 | |||
XM_005252997.1 | XP_005253054.1 | 4 | 1129 | |||
XM_017017953.1 | XP_016873442.1 | 4 | 1185 | |||
GenBank mRNA | AK027362.1 | - | 2 | 270 | ||
AK092290.1 | - | 4 | 486 | |||
AK127264.1 | - | 2 | 865 | |||
AK309240.1 | - | 3 | 533 |