Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPY19L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172774.1 | NP_001166245.1 | 19 | 4371 | ||
NM_207325.2 | NP_997208.2 | 19 | 4397 | |||
XM_006723045.1 | XP_006723108.1 | 16 | 3913 | |||
XM_011526526.2 | XP_011524828.1 | 19 | 4352 | |||
XM_017026366.1 | XP_016881855.1 | 18 | 4286 | |||
XM_017026367.1 | XP_016881856.1 | 19 | 4311 | |||
GenBank mRNA | AK024775.1 | - | 1 | 337 | ||
AK125846.1 | - | 3 | 2515 | |||
AK126757.1 | - | 19 | 4388 | |||
AL833713.1 | - | 9 | 4350 | |||
CR749459.1 | - | 13 | 3567 |