Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207327.4 | NP_997210.3 | 3 | 172 | ||
XM_011529960.1 | XP_011528262.1 | 3 | 555 | |||
XM_011529962.2 | XP_011528264.1 | 3 | 163 | |||
XM_011529963.1 | XP_011528265.1 | 3 | 553 | |||
XM_011529964.1 | XP_011528266.1 | 3 | 553 | |||
XM_011529965.2 | XP_011528267.1 | 3 | 488 | |||
GenBank mRNA | BC036462.1 | - | 3 | 170 | ||
BC045796.2 | - | 2 | 102 | |||
BC067871.1 | - | 3 | 172 | 30338205 | ||
BX648331.1 | - | 3 | 180 |