Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207523.2 | NP_997406.1 | 5 | 526 | ||
GenBank mRNA | AK028387.1 | - | 5 | 499 | ||
AK152095.1 | - | 5 | 499 | |||
BC016558.1 | - | 4 | 465 | |||
BC026656.1 | - | 5 | 483 | |||
BC029892.1 | - | 5 | 485 | 4163533 | ||
BC086883.1 | - | 5 | 501 | 6756980 | ||
BC086884.1 | - | 5 | 499 | 30122733 | ||
BC096637.1 | - | 5 | 484 | |||
BY585987.1 | - | 4 | 367 |