Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNASE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005223.3 | NP_005214.2 | 3, 4 | 2352 | ||
XM_005255148.3 | XP_005255205.1 | 6, 7 | 1263 | |||
XM_011522393.2 | XP_011520695.1 | 6, 7 | 1262 | |||
XM_017022992.1 | XP_016878481.1 | 6, 7 | 1264 | |||
XM_017022993.1 | XP_016878482.1 | 7, 8 | 1869 | |||
XM_017022994.1 | XP_016878483.1 | 8, 9 | 2035 | |||
XM_017022995.1 | XP_016878484.1 | 6, 7 | 1069 | |||
XM_017022996.1 | XP_016878485.1 | 6, 7 | 1015 | |||
XM_017022997.1 | XP_016878486.1 | 7, 8 | 1683 | |||
XM_017022998.1 | XP_016878487.1 | 5, 6 | 1968 | |||
XM_017022999.1 | XP_016878488.1 | 4, 5 | 1006 | |||
XM_017023003.1 | XP_016878492.1 | 3, 4 | 1171 | |||
XM_017023007.1 | XP_016878496.1 | 6, 7 | 1263 | |||
GenBank mRNA | AB188151.1 | - | 4, 5 | 431 | ||
AB188152.1 | - | 3, 4 | 485 | |||
AJ298844.1 | - | 3, 4 | 1307 | |||
BC029437.1 | - | 3, 4 | 373 | 4594247 | ||
M55983.1 | - | 3, 4 | 404 |