Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS37C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017966.4 | NP_060436.4 | 5 | 2122 | ||
XM_005274077.3 | XP_005274134.1 | 5 | 2163 | |||
XM_017017955.1 | XP_016873444.1 | 2 | 1832 | |||
GenBank mRNA | AK000854.1 | - | 1 | 710 | ||
AK302384.1 | - | 6 | 1142 | |||
AL834261.1 | - | 5 | 2130 | |||
BC005805.2 | - | 1 | 749 | |||
BC062604.1 | - | 1 | 845 | |||
BC141827.1 | - | 5 | 2085 | 40148285 | ||
BC142702.1 | - | 5 | 2085 | 40148798 | ||
CR457250.1 | - | 1 | 389 |