Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IWS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017969.2 | NP_060439.2 | 2 | 401 | ||
XM_005263707.3 | XP_005263764.1 | 2 | 237 | |||
XM_006712626.3 | XP_006712689.1 | 2 | 315 | |||
XM_011511450.1 | XP_011509752.1 | 2 | 315 | |||
XM_011511452.1 | XP_011509754.1 | 2 | 315 | |||
XM_017004467.1 | XP_016859956.1 | 2 | 402 | |||
XM_017004468.1 | XP_016859957.1 | 2 | 171 | |||
GenBank mRNA | AK000868.1 | - | 2 | 349 | ||
AK027561.1 | - | 2 | 358 | |||
AK294669.1 | - | 2 | 379 | |||
AK295970.1 | - | 2 | 164 | |||
AK296848.1 | - | 2 | 353 | |||
AL834178.1 | - | 2 | 349 | |||
BC065279.1 | - | 2 | 160 | |||
BC110536.1 | - | 2 | 231 | 40035173 | ||
BC110537.1 | - | 2 | 231 | 40035174 | ||
DA268882.1 | - | 2 | 402 |