Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000454.1 | NP_001000454.1 | 1 | 436 |
Gene Symbol: LOC100909736 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750485.1 | XP_003750533.1 | 247 |
Gene Symbol: LOC100912507 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762440.1 | XP_008760662.1 | 1 | 436 |
Gene Symbol: LOC103692018 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762660.1 | XP_008760882.1 | 1 | 436 |