Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr361 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000556.1 | NP_001000556.1 | 1 | 483 |
Gene Symbol: LOC103691301 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760292.1 | XP_008758514.1 | 1 | 483 |