Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000964.1 | NP_001000964.1 | 118 |
Gene Symbol: LOC100910899 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750473.2 | XP_003750521.1 | 1 | 118 |
Gene Symbol: LOC100911845 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749655.1 | XP_003749703.1 | 1 | 118 |