Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127603.1 | NP_001121075.1 | 5 | 511 | ||
NM_017881.2 | NP_060351.1 | 5 | 511 | |||
NR_023352.1 | 6 | 768 | ||||
XM_006717162.1 | XP_006717225.1 | 6 | 796 | |||
XM_006717163.2 | XP_006717226.1 | 6 | 831 | |||
XM_011518817.1 | XP_011517119.1 | 5 | 621 | |||
XM_011518818.1 | XP_011517120.1 | 5 | 1143 | |||
XM_011518819.2 | XP_011517121.1 | 5 | 1143 | |||
XM_017014868.1 | XP_016870357.1 | 7 | 1117 | |||
XM_017014869.1 | XP_016870358.1 | 6 | 1424 | |||
XM_017014870.1 | XP_016870359.1 | 5 | 1167 | |||
XM_017014871.1 | XP_016870360.1 | 6 | 796 | |||
XM_017014872.1 | XP_016870361.1 | 7 | 1117 | |||
XM_017014873.1 | XP_016870362.1 | 7 | 1088 | |||
XM_017014874.1 | XP_016870363.1 | 6 | 831 | |||
GenBank mRNA | AK000566.1 | - | 5 | 476 | ||
AK023413.1 | - | 5 | 503 | |||
AK097144.1 | - | 6 | 741 | |||
AY611480.1 | - | 4 | 266 | |||
BC001366.2 | - | 5 | 419 | 3051165 | ||
BC036804.1 | - | 5 | 511 | 5727308 | ||
CB135264.1 | - | 5 | 511 |