Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009941.2 | NP_001009941.1 | 7 | 1622 | ||
NM_001009943.2 | NP_001009943.1 | 6 | 1543 | |||
NM_019046.2 | NP_061919.1 | 7 | 1622 | |||
GenBank mRNA | AL137614.1 | - | 4 | 477 | ||
BC062346.1 | - | 7 | 1108 | 3845142 |