Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other C1ORF159 SIRNA Assays ›
Gene Symbol:
C1orf159
Gene Name:
chromosome 1 open reading frame 159
Gene Aliases:
-
Chromosome Location:
Chr.1: 1081811 - 1116356 on Build GRCh38
UniGene ID:
Hs.235095
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
25818
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: C1orf159
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017891.4
NP_060361.4
10
1641
XM_005244765.2
XP_005244822.1
10
1817
XM_006710717.2
XP_006710780.1
11
2173
XM_006710718.2
XP_006710781.1
10
2101
XM_011541651.2
XP_011539953.1
11
2088
XM_011541652.2
XP_011539954.1
13
3085
XM_011541653.2
XP_011539955.1
12
2182
XM_011541655.2
XP_011539957.1
11
3126
XM_011541656.2
XP_011539958.1
11
2656
XM_011541658.2
XP_011539960.1
11
2843
XM_011541659.2
XP_011539961.1
11
1957
XM_011541660.2
XP_011539962.1
11
2492
XM_011541661.2
XP_011539963.1
11
1938
XM_011541662.2
XP_011539964.1
12
2281
XM_011541663.2
XP_011539965.1
12
1966
XM_011541665.2
XP_011539967.1
11
1858
XM_017001572.1
XP_016857061.1
12
3272
XM_017001573.1
XP_016857062.1
11
2974
XM_017001574.1
XP_016857063.1
11
3178
XM_017001575.1
XP_016857064.1
12
2136
XM_017001576.1
XP_016857065.1
12
3228
XM_017001577.1
XP_016857066.1
10
2045
XM_017001578.1
XP_016857067.1
10
2504
XM_017001579.1
XP_016857068.1
9
2744
XM_017001580.1
XP_016857069.1
12
2834
GenBank mRNA
AK000591.1
-
10
1376
AK024796.1
-
2
746
AK054793.1
-
10
2292
AK057368.1
-
12
1966
AK124054.1
-
12
1643
AK128271.1
-
3
2859
AK128434.1
-
10
2758
AY358490.1
-
11
1552
BC008788.2
-
9
1474
3627081
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-9vv5m:80/100.66.78.111:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline