Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039671.2 | NP_001034760.1 | 8 | 2658 | ||
NM_001039672.2 | NP_001034761.1 | 8 | 2676 | |||
NM_001039673.2 | NP_001034762.1 | 8 | 2667 | |||
NM_001145461.1 | NP_001138933.1 | 8 | 2652 | |||
NM_001145462.1 | NP_001138934.1 | 8 | 2628 | |||
NM_001145463.1 | NP_001138935.1 | 9 | 2744 | |||
GenBank mRNA | AK027130.1 | - | 1 | 1351 | ||
BC025257.1 | - | 1 | 1287 |