Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024148.1 | NP_001019319.1 | 7 | 2306 | ||
GenBank mRNA | BC048920.1 | - | 6 | 2308 | ||
BC058786.1 | - | 7 | 2306 | 6310060 |
Gene Symbol: Ugt2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053184.2 | NP_444414.2 | 7 | 2337 | ||
GenBank mRNA | AK140757.1 | - | 7 | 2337 | ||
BC048926.1 | - | 6 | 2325 | 6314878 |