Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114108.1 | NP_001107580.1 | 4, 5 | 971 | ||
NM_017904.3 | NP_060374.2 | 4, 5 | 971 | |||
XM_011541671.2 | XP_011539973.1 | 4, 5 | 2082 | |||
XM_017001582.1 | XP_016857071.1 | 4, 5 | 2105 | |||
GenBank mRNA | AK000626.1 | - | 4, 5 | 901 |