Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HES2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019089.4 | NP_061962.2 | 4 | 2768 | ||
GenBank mRNA | AK023754.1 | - | 1 | 1953 | ||
AK091122.1 | - | 4 | 2768 |