Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003909.4 | NP_001003909.2 | 20 | 6362 | ||
NM_001289527.1 | NP_001276456.1 | 20 | 6380 | |||
NM_001289528.1 | NP_001276457.1 | 19 | 6244 | |||
NM_001289529.1 | NP_001276458.1 | 20 | 5989 | |||
GenBank mRNA | AK010017.1 | - | 1 | 1432 | ||
AK129343.1 | - | 20 | 6166 | |||
BC079620.1 | - | 9 | 4051 | |||
CN526058.1 | - | 1 | 240 |