Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017909.3 | NP_060379.2 | 2 | 562 | ||
XM_017010988.1 | XP_016866477.1 | 2 | 535 | |||
GenBank mRNA | AK000634.1 | - | 2 | 505 | ||
AK292339.1 | - | 2 | 537 | |||
BC012081.1 | - | 1 | 399 | |||
BC106065.1 | - | 2 | 524 | 6165385 | ||
BC119683.1 | - | 2 | 427 | 40116343 |