Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165136.1 | NP_001158608.1 | 20 | 2715 | ||
NM_017912.3 | NP_060382.3 | 21 | 2823 | |||
XM_005263083.3 | XP_005263140.1 | 21 | 3020 | |||
XM_011532053.2 | XP_011530355.1 | 20 | 2914 | |||
XM_017008333.1 | XP_016863822.1 | 20 | 2381 | |||
GenBank mRNA | AF336798.1 | - | 21 | 2823 | ||
AK000644.1 | - | 8 | 911 | |||
AK097168.1 | - | 15 | 1638 | |||
AK225506.1 | - | 8 | 911 | |||
AK295832.1 | - | 21 | 2733 | |||
AL833664.1 | - | 10 | 1151 | |||
AY653201.1 | - | 20 | 2715 | |||
AY653202.1 | - | 20 | 2627 | |||
AY653203.1 | - | 18 | 2408 | |||
BC035775.1 | - | 7 | 763 | |||
BC042047.1 | - | 21 | 2699 | 5531519 | ||
BX647121.1 | - | 21 | 2499 |