Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC37L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017913.3 | NP_060383.2 | 5 | 938 | ||
GenBank mRNA | AK000497.1 | - | 5 | 933 | ||
AK000646.1 | - | 5 | 822 | |||
AK225510.1 | - | 5 | 822 | |||
BC014133.2 | - | 5 | 758 | 4622495 |