Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008887.1 | NP_001008887.1 | 1 | 103 | ||
XM_006249631.2 | XP_006249693.1 | 1 | 142 |
Gene Symbol: Serpinb3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249640.2 | XP_006249702.1 | 2 | 215 |