Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008889.1 | NP_001008889.1 | 2 | 242 | ||
GenBank mRNA | BC107904.1 | - | 2 | 735 | 7454897 |