Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URGCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077663.2 | NP_001071131.1 | 6 | 717 | ||
NM_001077664.2 | NP_001071132.1 | 6 | 489 | |||
NM_001290075.1 | NP_001277004.1 | 6 | 712 | |||
NM_001290076.1 | NP_001277005.1 | 7 | 611 | |||
NM_017920.4 | NP_060390.3 | 5 | 690 | |||
GenBank mRNA | AB040940.1 | - | 7 | 1014 | ||
AK302459.1 | - | 7 | 611 | |||
AK314339.1 | - | 5 | 236 | |||
AL832134.1 | - | 4 | 2330 | |||
AY078404.1 | - | 5 | 209 | |||
BC018426.1 | - | 5 | 211 | 4213413 | ||
BG722670.1 | - | 6 | 467 | |||
BX640797.1 | - | 6 | 252 | |||
BX648505.1 | - | 6 | 257 | |||
CD628752.1 | - | 6 | 483 | |||
CR749398.1 | - | 6 | 269 | |||
DA240479.1 | - | 5 | 255 | |||
DA316326.1 | - | 6 | 273 | |||
DC336174.1 | - | 6 | 489 |