Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAA30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011713.2 | NP_001011713.2 | 2 | 825 | ||
GenBank mRNA | AK092674.1 | - | 2 | 825 | ||
AK304257.1 | - | 2 | 429 | |||
BC118589.1 | - | 2 | 780 | 40085064 | ||
BC122557.1 | - | 3 | 669 | 40085054 |