Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glrx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013034.1 | NP_001013052.1 | 5 | 488 | ||
XM_006249955.1 | XP_006250017.1 | 4 | 527 | |||
XM_006249956.2 | XP_006250018.1 | 4 | 550 | |||
XM_006249957.2 | XP_006250019.1 | 4 | 349 | |||
XM_006249958.2 | XP_006250020.1 | 4 | 417 | |||
XM_008769523.1 | XP_008767745.1 | 5 | 485 | |||
XM_008769524.1 | XP_008767746.1 | 4 | 377 | |||
XM_008769525.1 | XP_008767747.1 | 4 | 433 | |||
XM_008769526.1 | XP_008767748.1 | 4 | 417 | |||
GenBank mRNA | BC079292.1 | - | 5 | 488 | 7134405 | |
FQ213555.1 | - | 4 | 293 |