Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glrx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013034.1 | NP_001013052.1 | 8 | 1315 | ||
GenBank mRNA | BC079292.1 | - | 8 | 1315 | 7134405 |