Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkaa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013367.3 | NP_001013385.3 | 7 | 1108 | ||
GenBank mRNA | AK147811.1 | - | 7 | 1085 | ||
AK160612.1 | - | 7 | 1089 | |||
AY885266.1 | - | 7 | 1067 | |||
BC086695.1 | - | 7 | 1078 | 30691866 | ||
CB245934.1 | - | 2 | 404 |