Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017928.3 | NP_001017928.1 | 2 | 78 | ||
NM_001308326.1 | NP_001295255.1 | 2 | 104 | |||
XM_011512410.2 | XP_011510712.1 | 2 | 56 | |||
XM_017005697.1 | XP_016861186.1 | 2 | 128 | |||
GenBank mRNA | BC054051.1 | - | 2 | 69 | ||
BC062725.1 | - | 2 | 78 | 6495960 | ||
BC109378.1 | - | 2 | 62 | |||
CD683826.1 | - | 2 | 100 | |||
DA025111.1 | - | 2 | 100 |